Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083895.3 | NP_001077364.2 | 9 | 1551 | ||
NM_026838.4 | NP_081114.2 | 10 | 1603 | |||
GenBank mRNA | AF393640.1 | - | 8 | 1121 | ||
AK136935.1 | - | 9 | 1406 | |||
BC028307.1 | - | 10 | 1332 | 3488526 | ||
BQ560499.1 | - | 2 | 126 |