Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199465.2 | NP_955759.2 | 11 | 1572 | ||
GenBank mRNA | AK135919.1 | - | 2 | 260 | ||
AK162232.1 | - | 6 | 1014 | |||
AK162924.1 | - | 3 | 484 | |||
BC052878.1 | - | 11 | 1519 | 30046316 | ||
HQ179982.1 | - | 11 | 1606 | |||
HQ179983.1 | - | 9 | 1375 | |||
HQ179984.1 | - | 8 | 1204 |