Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026845.4 | NP_081121.1 | 4 | 511 | ||
GenBank mRNA | AK004331.1 | - | 4 | 512 | ||
AK150643.1 | - | 4 | 488 | |||
AK151572.1 | - | 4 | 488 | |||
AK217140.1 | - | 3 | 330 | |||
BC037131.1 | - | 4 | 491 | |||
BC051386.1 | - | 4 | 478 | |||
BC058369.1 | - | 4 | 470 | 5715252 | ||
BC147647.1 | - | 4 | 596 | 9056637 | ||
BC147649.1 | - | 4 | 596 | 9056639 |