Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175134.4 | NP_780343.1 | 4 | 607 | ||
GenBank mRNA | AB076382.1 | - | 4 | 602 | ||
AK079270.1 | - | 2 | 166 | |||
AK088304.1 | - | 4 | 568 | |||
AK148049.1 | - | 4 | 568 | |||
BC035553.1 | - | 4 | 2185 | |||
BC086659.1 | - | 4 | 574 | 30607272 | ||
BC094595.1 | - | 4 | 570 | 5707829 | ||
BQ715954.1 | - | 4 | 607 |