Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278218.1 | NP_001265147.1 | 9 | 3252 | ||
NM_001278219.1 | NP_001265148.1 | 9 | 3212 | |||
NM_001278220.1 | NP_001265149.1 | 8 | 3095 | |||
NM_026866.3 | NP_081142.3 | 10 | 3340 | |||
GenBank mRNA | AK135755.1 | - | 1 | 1147 | ||
AK138276.1 | - | 10 | 3297 | |||
AK170113.1 | - | 9 | 3207 | |||
AY144589.1 | - | 8 | 3050 | |||
AY150577.1 | - | 8 | 2999 | |||
AY150698.1 | - | 10 | 3232 | |||
BC043102.1 | - | 9 | 3213 | 6406376 | ||
BC059225.1 | - | 9 | 3213 | 6406376 |