Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278218.1 | NP_001265147.1 | 9 | 4891 | ||
NM_001278219.1 | NP_001265148.1 | 9 | 4851 | |||
NM_001278220.1 | NP_001265149.1 | 8 | 4734 | |||
NM_026866.3 | NP_081142.3 | 10 | 4979 | |||
GenBank mRNA | AK138276.1 | - | 10 | 4936 | ||
AK170113.1 | - | 9 | 4846 | |||
AY150577.1 | - | 8 | 4638 | |||
AY150698.1 | - | 10 | 4871 | |||
BB732349.1 | - | 1 | 385 | |||
BC043102.1 | - | 9 | 4852 | 6406376 | ||
BC059225.1 | - | 9 | 4852 | 6406376 | ||
BX526271.1 | - | 1 | 460 |