Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026872.1 | NP_081148.1 | 12 | 1102 | ||
GenBank mRNA | AF276965.1 | - | 11 | 1006 | ||
AK204028.1 | - | 4 | 253 | |||
AK214539.1 | - | 2 | 194 | |||
BC007179.1 | - | 12 | 1102 | 3496608 |