Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026909.2 | NP_081185.1 | 3 | 217 | ||
GenBank mRNA | AK007329.1 | - | 2 | 169 | ||
AK154641.1 | - | 3 | 303 | |||
BC018221.1 | - | 3 | 217 | 4215606 |