Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022885.2 | NP_075023.2 | 12 | 1797 | ||
GenBank mRNA | AF233321.1 | - | 5 | 816 | ||
AF461761.1 | - | 12 | 1696 | |||
AK007424.1 | - | 3 | 317 | |||
AK051183.1 | - | 12 | 1797 | |||
AK088004.1 | - | 11 | 1541 | |||
AK090176.1 | - | 12 | 1706 | |||
AK166006.1 | - | 12 | 1607 | |||
BC029033.1 | - | 9 | 1239 | |||
BC029217.1 | - | 5 | 649 | |||
BC033452.1 | - | 12 | 1696 | 3156622 |