Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810013L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081400.3 | NP_001074869.1 | 4 | 965 | ||
GenBank mRNA | AK007485.1 | - | 4 | 965 | ||
BC116778.1 | - | 4 | 948 | 40126097 | ||
BC116780.1 | - | 4 | 948 | 40126099 |