Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026936.3 | NP_081212.1 | 5 | 600 | ||
GenBank mRNA | AK007567.1 | - | 5 | 602 | ||
AK028379.1 | - | 5 | 599 | |||
AK044817.1 | - | 5 | 599 | |||
AK052432.1 | - | 5 | 601 | |||
AK077624.1 | - | 5 | 601 | |||
AK087756.1 | - | 5 | 604 | |||
AK139484.1 | - | 5 | 608 | |||
AK154304.1 | - | 5 | 603 | |||
AK154532.1 | - | 5 | 601 | |||
AK170898.1 | - | 5 | 601 | |||
AK214867.1 | - | 3 | 190 | |||
BC027191.1 | - | 6 | 532 | 4223847 |