Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810030O07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175141.5 | NP_780350.1 | 5 | 706 | ||
GenBank mRNA | AK007661.1 | - | 5 | 734 | ||
AK078166.1 | - | 5 | 707 | |||
AK146041.1 | - | 5 | 728 | |||
AK169085.1 | - | 5 | 713 | |||
AK178929.1 | - | 4 | 411 | |||
AK181549.1 | - | 4 | 411 | |||
BC038642.1 | - | 5 | 694 | |||
BC051146.1 | - | 5 | 718 | |||
BC137660.1 | - | 5 | 799 | 8860679 |