Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026965.2 | NP_081241.1 | 7 | 717 | ||
GenBank mRNA | AK007659.1 | - | 7 | 715 | ||
AK054334.1 | - | 7 | 718 | |||
BC019467.1 | - | 6 | 775 | |||
BC049670.1 | - | 7 | 712 | 6535487 | ||
CT010388.1 | - | 7 | 679 |