Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026977.2 | NP_081253.1 | 6 | 727 | ||
GenBank mRNA | AK029090.1 | - | 6 | 727 | ||
AK145962.1 | - | 7 | 974 | |||
AK159252.1 | - | 449 | ||||
AK169779.1 | - | 6 | 999 | |||
AK177396.1 | - | 2 | 50 | |||
AK177413.1 | - | 2 | 50 | |||
AK200521.1 | - | 194 | ||||
AK204709.1 | - | 2 | 50 | |||
BC013714.1 | - | 5 | 500 | |||
BC028815.1 | - | 6 | 712 | 5126467 |