Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040396.2 | NP_001035486.2 | 4 | 512 | ||
GenBank mRNA | AK013022.1 | - | 4 | 476 | ||
AK032838.1 | - | 4 | 489 | |||
AK047958.1 | - | 4 | 490 | |||
AK146778.1 | - | 4 | 400 | |||
BC019970.1 | - | 4 | 460 | 4458171 | ||
BC038867.1 | - | 4 | 388 | 1547418 | ||
BC043068.1 | - | 398 | ||||
BC132453.1 | - | 4 | 478 | 40130730 | ||
BC138424.1 | - | 4 | 478 | 8861444 |