Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040396.2 | NP_001035486.2 | 3 | 381 | ||
GenBank mRNA | AK013022.1 | - | 3 | 345 | ||
AK032838.1 | - | 3 | 358 | |||
AK047958.1 | - | 3 | 359 | |||
AK146778.1 | - | 3 | 269 | |||
BC019970.1 | - | 3 | 329 | 4458171 | ||
BC038867.1 | - | 3 | 257 | 1547418 | ||
BC043068.1 | - | 267 | ||||
BC132453.1 | - | 3 | 347 | 40130730 | ||
BC138424.1 | - | 3 | 347 | 8861444 | ||
CJ133950.1 | - | 3 | 382 |