Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027016.2 | NP_081292.1 | 8 | 1546 | ||
GenBank mRNA | AK075621.1 | - | 1 | 54 | ||
AK088093.1 | - | 8 | 1095 | |||
AK090078.1 | - | 8 | 1100 | |||
BC023720.1 | - | 5 | 761 | |||
BC053504.1 | - | 8 | 1084 | |||
BC067202.1 | - | 8 | 1081 | 6332701 | ||
BC071243.1 | - | 8 | 1095 |