Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081347.1 | NP_001074816.1 | 6, 7 | 1589 | ||
NM_001252636.1 | NP_001239565.1 | 6, 7 | 1572 | |||
NM_001252637.1 | NP_001239566.1 | 6, 7 | 1693 | |||
NM_001252638.1 | NP_001239567.1 | 7, 8 | 1752 | |||
GenBank mRNA | AK005770.1 | - | 6, 7 | 1572 | ||
AK034826.1 | - | 6, 7 | 1589 | |||
AK078306.1 | - | 5, 6 | 1526 | |||
AK173015.1 | - | 1, 2 | 953 | |||
BC099973.1 | - | 6, 7 | 1570 | 6432524 | ||
BC137646.1 | - | 5, 6 | 1523 | 8860665 | ||
BC137647.1 | - | 5, 6 | 1523 | 8860666 | ||
BC144800.1 | - | 6, 7 | 1582 | 9053348 |