Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027052.3 | NP_081328.2 | 14 | 1532 | ||
GenBank mRNA | AB055004.1 | - | 14 | 1508 | ||
AK005887.1 | - | 1 | 59 | |||
AK051023.1 | - | 14 | 1533 | |||
AK164292.1 | - | 14 | 1475 | |||
AK167539.1 | - | 14 | 1514 | |||
AK169349.1 | - | 13 | 1521 | |||
AY027919.1 | - | 14 | 1571 | |||
BC024072.1 | - | 14 | 1457 | 5097293 | ||
BC024123.1 | - | 14 | 1489 | 5098067 | ||
BC031717.1 | - | 14 | 1450 | 4195569 |