Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166494.1 | NP_001159966.1 | 6 | 1125 | ||
NM_001166495.1 | NP_001159967.1 | 7 | 914 | |||
NM_027061.2 | NP_081337.2 | 7 | 1005 | |||
NM_199419.3 | NP_955451.2 | 6 | 786 | |||
GenBank mRNA | AK006058.1 | - | 6 | 1073 | ||
AY251601.1 | - | 7 | 950 | |||
AY251602.1 | - | 6 | 731 | |||
BC050986.1 | - | 7 | 881 | |||
BC089476.1 | - | 6 | 786 | 6708062 |