Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023514.4 | NP_076003.3 | 2 | 252 | ||
GenBank mRNA | AK009082.1 | - | 2 | 202 | ||
AK031497.1 | - | 2 | 201 | |||
AK049465.1 | - | 2 | 210 | |||
AK144656.1 | - | 2 | 210 | |||
AK150985.1 | - | 2 | 210 | |||
AK214977.1 | - | 2 | 90 | |||
BC083159.1 | - | 1 | 13 | |||
BC116437.2 | - | 2 | 202 | 40104493 | ||
BY145802.1 | - | 2 | 252 |