Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181588.3 | NP_853619.1 | 2 | 231 | ||
GenBank mRNA | AK009374.1 | - | 1 | 221 | ||
BC024580.1 | - | 2 | 255 | 5064316 |