Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181588.3 | NP_853619.1 | 4, 5 | 514 | ||
GenBank mRNA | AK009374.1 | - | 3, 4 | 506 | ||
BC024580.1 | - | 4, 5 | 538 | 5064316 |