Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181588.3 | NP_853619.1 | 6 | 750 | ||
GenBank mRNA | AK009374.1 | - | 5 | 742 | ||
BC024580.1 | - | 6 | 774 | 5064316 |