Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027127.2 | NP_081403.1 | 1 | 27 | ||
GenBank mRNA | AK009378.1 | - | 1 | 28 | ||
BC019664.1 | - | 1 | 26 | 3988837 |