Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027132.2 | NP_081408.2 | 6 | 1536 | ||
GenBank mRNA | AB087505.1 | - | 6 | 1434 | ||
AK043266.1 | - | 6 | 1434 | |||
BC145613.1 | - | 6 | 1396 | 9054179 | ||
BC150803.1 | - | 6 | 1396 | 9087714 |