Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145131.1 | NP_660113.1 | 25 | 2830 | ||
GenBank mRNA | AF513714.1 | - | 25 | 2830 | ||
AK009313.1 | - | 2 | 143 | |||
AK129292.1 | - | 25 | 2804 | |||
AK141277.1 | - | 25 | 2825 | |||
AK147837.1 | - | 25 | 2821 | |||
AK164235.1 | - | 25 | 2792 | |||
AK167426.1 | - | 25 | 2821 | |||
AK168335.1 | - | 24 | 2707 | |||
AY779273.1 | - | 25 | 2830 | |||
AY779274.1 | - | 25 | 2830 | |||
BC006917.1 | - | 15 | 1631 | 3601081 |