Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027156.3 | NP_081432.2 | 6 | 907 | ||
GenBank mRNA | AK141361.1 | - | 6 | 888 | ||
AK152599.1 | - | 6 | 875 | |||
BC023700.1 | - | 3 | 170 | |||
BC060646.1 | - | 6 | 884 | 6853826 |