Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175150.3 | NP_780359.2 | 2 | 624 | ||
GenBank mRNA | AK009878.1 | - | 2 | 552 | ||
AK046071.1 | - | 1 | 382 | |||
AK079517.1 | - | 2 | 571 | |||
AK160051.1 | - | 2 | 593 | |||
AK167701.1 | - | 2 | 532 | |||
AK167838.1 | - | 2 | 590 | |||
AK167977.1 | - | 2 | 589 | |||
BC029015.1 | - | 1 | 360 | |||
BC062180.1 | - | 2 | 624 | 6336176 | ||
BC094275.1 | - | 2 | 460 |