Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181395.2 | NP_852060.2 | 9 | 957 | ||
GenBank mRNA | AK054255.1 | - | 8 | 872 | ||
AK122223.1 | - | 9 | 727 | |||
AK142872.1 | - | 9 | 957 | |||
AK163597.1 | - | 9 | 944 | |||
BC150789.1 | - | 9 | 952 | 9087700 |