Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181395.2 | NP_852060.2 | 7 | 711 | ||
GenBank mRNA | AK054255.1 | - | 6 | 626 | ||
AK122223.1 | - | 7 | 481 | |||
AK142872.1 | - | 7 | 711 | |||
AK163597.1 | - | 7 | 698 | |||
BC150789.1 | - | 7 | 706 | 9087700 |