Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178378.3 | NP_848465.1 | 4 | 475 | ||
GenBank mRNA | AK146422.1 | - | 2 | 213 | ||
BC049990.1 | - | 4 | 459 | 6310844 |