Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027184.1 | NP_081460.1 | 6 | 1092 | ||
GenBank mRNA | AK036084.1 | - | 6 | 860 | ||
AK036978.1 | - | 6 | 1052 | |||
AK039460.1 | - | 1 | 36 | |||
AK172246.1 | - | 6 | 1184 | |||
BC052463.1 | - | 6 | 1092 | 6834748 |