Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177561.3 | NP_808229.1 | 3, 4 | 456 | ||
GenBank mRNA | AK029604.1 | - | 2, 3 | 308 | ||
AK145298.1 | - | 3, 4 | 476 | |||
BC030888.1 | - | 1, 2 | 223 | |||
BC039916.1 | - | 3, 4 | 458 | 4950034 |