Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177561.3 | NP_808229.1 | 5, 6 | 756 | ||
GenBank mRNA | AK029604.1 | - | 4, 5 | 608 | ||
AK145298.1 | - | 5, 6 | 776 | |||
BC030888.1 | - | 3, 4 | 523 | |||
BC039916.1 | - | 5, 6 | 758 | 4950034 |