Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027191.2 | NP_081467.2 | 2 | 269 | ||
NM_028334.4 | NP_082610.1 | 2 | 287 | |||
GenBank mRNA | AK010370.1 | - | 2 | 213 | ||
AK012890.1 | - | 2 | 283 | |||
AK040463.1 | - | 2 | 270 | |||
AK043277.1 | - | 1 | 1324 | |||
AK165537.1 | - | 2 | 233 | |||
BC011102.1 | - | 2 | 253 | 3982076 | ||
BC032180.1 | - | 1 | 197 | 5151847 | ||
CJ072834.1 | - | 2 | 289 |