Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027193.2 | NP_081469.2 | 3, 4 | 388 | ||
GenBank mRNA | AK010475.1 | - | 3, 4 | 378 | ||
AK034652.1 | - | 398 | ||||
AK045127.1 | - | 3, 4 | 408 | |||
AK045548.1 | - | 3, 4 | 378 | |||
AK046024.1 | - | 3, 4 | 378 | |||
AK046753.1 | - | 3, 4 | 406 | |||
BC060372.1 | - | 3, 4 | 388 | 6391546 |