Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133716.3 | NP_598477.2 | 4, 5 | 808 | ||
GenBank mRNA | AK152757.1 | - | 4, 5 | 775 | ||
BC031581.1 | - | 4, 5 | 797 | |||
BC032217.1 | - | 4, 5 | 784 | |||
BC052413.1 | - | 4, 5 | 808 | 5720177 |