Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027211.2 | NP_081487.1 | 745 | |||
GenBank mRNA | AJ306451.1 | - | 745 | |||
AK007692.1 | - | 1 | 126 | |||
BC013521.1 | - | 715 | 4236501 |