Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159349.1 | NP_001152821.1 | 7 | 832 | ||
NM_027226.4 | NP_081502.2 | 8 | 934 | |||
GenBank mRNA | AJ344096.1 | - | 8 | 818 | ||
AK014447.1 | - | 8 | 905 | |||
AK029785.1 | - | 8 | 879 | |||
AK050448.1 | - | 8 | 893 | |||
AK050479.1 | - | 8 | 906 | |||
AK077184.1 | - | 9 | 842 | |||
AK139638.1 | - | 7 | 828 | |||
AK157602.1 | - | 8 | 906 | |||
AK161252.1 | - | 8 | 912 | |||
AK166032.1 | - | 8 | 906 | |||
AK170511.1 | - | 8 | 906 | |||
AK200270.1 | - | 1 | 103 | |||
BC010204.1 | - | 8 | 908 | 3158224 | ||
BC028253.1 | - | 8 | 891 | 3983533 |