Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178620.3 | NP_848735.1 | 13 | 1284 | ||
GenBank mRNA | AF015191.1 | - | 13 | 1250 | ||
AK031384.1 | - | 13 | 1479 | |||
AK032043.1 | - | 13 | 1480 | |||
AK191123.1 | - | 2 | 56 | |||
BC016209.1 | - | 14 | 1243 | |||
BC079583.1 | - | 13 | 1311 | |||
BC088988.1 | - | 13 | 1284 | 6418757 | ||
BC100547.1 | - | 13 | 3360 | |||
BC137970.1 | - | 13 | 1271 | 8860989 | ||
BC137971.1 | - | 13 | 1272 | 8860990 | ||
BC145068.1 | - | 13 | 1251 |