Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172402.3 | NP_765990.2 | 4 | 691 | ||
GenBank mRNA | AK031187.1 | - | 4 | 496 | ||
AK145178.1 | - | 4 | 651 | |||
AK167501.1 | - | 4 | 501 | |||
AK170643.1 | - | 4 | 660 | |||
AK172029.1 | - | 4 | 659 | |||
AK172597.1 | - | 4 | 649 | |||
BC031874.1 | - | 4 | 625 | 5149984 |