Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023538.2 | NP_076027.1 | 6 | 408 | ||
GenBank mRNA | AJ401619.1 | - | 6 | 381 | ||
AK011715.1 | - | 6 | 404 | |||
AK076224.1 | - | 5 | 371 | |||
AY772469.1 | - | 5 | 349 | |||
BC019145.1 | - | 6 | 402 | 5051535 | ||
BC093525.1 | - | 6 | 378 | 5151866 |