Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175154.3 | NP_780363.1 | 1, 2 | 263 | ||
GenBank mRNA | AK082521.1 | - | 1, 2 | 154 |