Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 6 | 962 | ||
GenBank mRNA | AK008408.1 | - | 6 | 847 | ||
AK077065.1 | - | 6 | 965 | |||
AK150071.1 | - | 6 | 865 | |||
AK159364.1 | - | 6 | 889 | |||
AK161096.1 | - | 6 | 844 | |||
AK170171.1 | - | 6 | 865 | |||
AK170697.1 | - | 6 | 866 | |||
AK212809.1 | - | 2 | 105 | |||
BC018367.1 | - | 6 | 814 | 2648510 | ||
BC018491.1 | - | 6 | 842 | 2651602 | ||
BC026136.1 | - | 5 | 438 |