Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133718.4 | NP_598479.1 | 1 | 429 | ||
GenBank mRNA | AK008408.1 | - | 1 | 315 | ||
AK077065.1 | - | 1 | 432 | |||
AK150071.1 | - | 1 | 332 | |||
AK159364.1 | - | 1 | 356 | |||
AK161096.1 | - | 1 | 311 | |||
AK170171.1 | - | 1 | 332 | |||
AK170697.1 | - | 1 | 333 | |||
AK178013.1 | - | 1 | 69 | |||
AK192822.1 | - | 1 | 69 | |||
AK200429.1 | - | 1 | 93 | |||
BC018367.1 | - | 1 | 281 | 2648510 | ||
BC018491.1 | - | 1 | 309 | 2651602 |