Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130513.1 | NP_001123985.1 | 10 | 1497 | ||
NM_027286.4 | NP_081562.2 | 9 | 1349 | |||
GenBank mRNA | AB053181.1 | - | 9 | 1353 | ||
AK135357.1 | - | 4 | 531 | |||
AK141604.1 | - | 9 | 1198 | |||
BC026801.1 | - | 9 | 1342 | 4236529 | ||
EF408739.1 | - | 10 | 1458 | |||
EF408740.1 | - | 10 | 1458 | |||
GQ262785.1 | - | 9 | 1167 |