Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197993.2 | NP_932110.1 | 9 | 2284 | ||
GenBank mRNA | AK215597.1 | - | 1 | 260 | ||
BC023900.1 | - | 9 | 2261 | 5321265 | ||
BC025499.1 | - | 9 | 2308 | 5320398 |