Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027315.4 | NP_081591.3 | 11 | 1200 | ||
GenBank mRNA | AK009324.1 | - | 10 | 782 | ||
AK146707.1 | - | 11 | 1013 | |||
AY112699.1 | - | 11 | 1193 | |||
AY945937.1 | - | 11 | 1097 | |||
BC030044.1 | - | 8 | 525 | |||
BC051487.1 | - | 4 | 197 | |||
BC060966.1 | - | 9 | 593 | |||
BC082275.1 | - | 11 | 1023 | 6310111 |