Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027338.1 | NP_081614.1 | 12 | 946 | ||
GenBank mRNA | AK005810.1 | - | 1 | 113 | ||
AK150754.1 | - | 12 | 967 | |||
AK151035.1 | - | 12 | 967 | |||
AK156241.1 | - | 961 | ||||
AK161865.1 | - | 998 | ||||
BC010811.1 | - | 12 | 946 | 4210911 |